Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERTAD4
Entrez Gene ID
56256
Gene Name
SERTA domain containing 4
Gene Aliases
DJ667H12.2
Chromosome Location
Chr.1: 210232650 - 210247734 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005273193.4 XP_005273250.1 4 - 5 800 89

View Full Product Details