The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304271.1 | NP_001291200.1 | 1 - 2 | 426 | 63 |
NM_001304272.1 | NP_001291201.1 | 1 - 2 | 426 | 63 | |
NM_001304273.1 | NP_001291202.1 | 1 - 2 | 426 | 63 | |
NM_024585.3 | NP_078861.1 | 1 - 2 | 426 | 63 | |
GenBank mRNA | AK025813.1 | - | 1 - 2 | 234 | 63 |
AK300997.1 | - | 1 - 2 | 426 | 63 | |
BC011728.1 | - | 1 - 2 | 391 | 63 | |
BI826938.1 | - | 1 - 2 | 392 | 63 |