The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243108.1 | NP_001230037.1 | 17 - 18 | 1947 | 106 |
NM_002663.4 | NP_002654.3 | 17 - 18 | 1947 | 106 | |
XM_005256696.2 | XP_005256753.1 | 8 - 9 | 1007 | 106 | |
XM_017024764.1 | XP_016880253.1 | 17 - 18 | 1947 | 106 | |
GenBank mRNA | AF033850.1 | - | 17 - 18 | 1982 | 106 |
AF035483.1 | - | 16 - 17 | 1821 | 106 | |
AF038441.1 | - | 17 - 18 | 1905 | 106 | |
AK304246.1 | - | 17 - 18 | 1900 | 106 | |
AK314797.1 | - | 17 - 18 | 1865 | 106 | |
AY178289.1 | - | 8 - 9 | 912 | 106 | |
BC015033.1 | - | 17 - 18 | 1908 | 106 | |
BC056871.1 | - | 17 - 18 | 1856 | 106 |