The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142286.1 | NP_001135758.1 | 3 - 4 | 415 | 101 |
NM_024624.5 | NP_078900.1 | 2 - 3 | 328 | 101 | |
XM_011533107.2 | XP_011531409.1 | 1 - 2 | 1648 | 101 | |
XM_011533108.2 | XP_011531410.1 | 2 - 3 | 324 | 101 | |
XM_017004913.1 | XP_016860402.1 | 2 - 3 | 327 | 101 | |
XM_017004914.1 | XP_016860403.1 | 3 - 4 | 417 | 101 | |
XM_017004915.1 | XP_016860404.1 | 1 - 2 | 1648 | 101 | |
XM_017004916.1 | XP_016860405.1 | 2 - 3 | 324 | 101 | |
GenBank mRNA | AJ310551.1 | - | 1 - 2 | 119 | 101 |
AK092853.1 | - | 1 - 2 | 585 | 101 | |
AK098325.1 | - | 1 - 2 | 58 | 101 | |
AK292421.1 | - | 5 - 6 | 521 | 101 | |
AL136544.1 | - | 3 - 4 | 389 | 101 | |
AL832979.1 | - | 2 - 3 | 291 | 101 | |
BC032675.1 | - | 3 - 4 | 370 | 101 | |
BC039828.1 | - | 2 - 3 | 312 | 101 |