The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024857.4 | NP_079133.3 | 16 - 17 | 4298 | 105 |
XM_011525269.2 | XP_011523571.1 | 16 - 17 | 4322 | 105 | |
XM_011525270.2 | XP_011523572.1 | 15 - 16 | 4145 | 105 | |
XM_011525274.2 | XP_011523576.1 | 11 - 12 | 1550 | 105 | |
XM_017025111.1 | XP_016880600.1 | 16 - 17 | 4297 | 105 | |
GenBank mRNA | AJ314648.2 | - | 16 - 17 | 4266 | 105 |
AY557611.1 | - | 16 - 17 | 4266 | 105 |