Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PUM2
Entrez Gene ID
23369
Gene Name
pumilio RNA binding family member 2
Gene Aliases
PUMH2, PUML2
Chromosome Location
Chr.2: 20248692 - 20352423 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282752.1 NP_001269681.1 18 - 19 3032 68
NM_001282790.1 NP_001269719.1 17 - 18 2658 68
NM_001282791.1 NP_001269720.1 18 - 19 2783 68
NM_015317.2 NP_056132.1 19 - 20 3141 68
XM_005262607.1 XP_005262664.1 19 - 20 3121 68
XM_005262609.1 XP_005262666.1 18 - 19 2884 68
XM_005262610.1 XP_005262667.1 18 - 19 2878 68
XM_006711973.1 XP_006712036.1 19 - 20 2918 68
XM_006711974.2 XP_006712037.1 19 - 20 2958 68
XM_006711975.2 XP_006712038.1 20 - 21 3082 68
XM_011532719.1 XP_011531021.1 21 - 22 3317 68
XM_011532720.2 XP_011531022.1 21 - 22 3326 68
XM_011532721.2 XP_011531023.1 20 - 21 3118 68
XM_011532723.1 XP_011531025.1 18 - 19 2748 68
XM_017003701.1 XP_016859190.1 19 - 20 2912 68
XM_017003702.1 XP_016859191.1 19 - 20 2952 68
XM_017003703.1 XP_016859192.1 20 - 21 3146 68
XM_017003704.1 XP_016859193.1 22 - 23 3464 68
XM_017003705.1 XP_016859194.1 22 - 23 3451 68
XM_017003706.1 XP_016859195.1 21 - 22 3243 68
XM_017003707.1 XP_016859196.1 18 - 19 2681 68
XM_017003708.1 XP_016859197.1 18 - 19 2914 68
XM_017003709.1 XP_016859198.1 19 - 20 2874 68
XM_017003710.1 XP_016859199.1 18 - 19 2724 68
XM_017003711.1 XP_016859200.1 18 - 19 2675 68
XM_017003712.1 XP_016859201.1 18 - 19 2743 68
XM_017003713.1 XP_016859202.1 19 - 20 2902 68
XM_017003714.1 XP_016859203.1 15 - 16 2521 68
XM_017003715.1 XP_016859204.1 17 - 18 2505 68
GenBank mRNA AF272350.1 - 15 - 16 2305 68
AF315591.1 - 18 - 19 2892 68
AK093847.1 - 19 - 20 2993 68
AK304198.1 - 18 - 19 2802 68
BC024218.2 - 7 - 8 1144 68
BC112046.1 - 18 - 19 2895 68
BC112048.1 - 18 - 19 2895 68
BC143550.1 - 17 - 18 2658 68
BC143552.1 - 18 - 19 2783 68
D87078.3 - 18 - 19 2869 68

View Full Product Details