The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001123384.1 | NP_001116856.1 | 4 - 5 | 3362 | 84 |
XM_005272619.3 | XP_005272676.1 | 4 - 5 | 3749 | 84 | |
XM_005272620.3 | XP_005272677.1 | 4 - 5 | 3749 | 84 | |
GenBank mRNA | AK074286.1 | - | 1 - 2 | 2352 | 84 |
BC114220.1 | - | 4 - 5 | 3362 | 84 |