The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001123383.1 | NP_001116855.1 | 12 - 13 | 4991 | 109 |
NM_001123384.1 | NP_001116856.1 | 11 - 12 | 4937 | 109 | |
NM_001123385.1 | NP_001116857.1 | 12 - 13 | 5022 | 109 | |
NM_017745.5 | NP_060215.4 | 12 - 13 | 4920 | 109 | |
XM_005272616.1 | XP_005272673.1 | 12 - 13 | 5154 | 109 | |
XM_005272618.3 | XP_005272675.1 | 12 - 13 | 4820 | 109 | |
XM_005272619.3 | XP_005272676.1 | 11 - 12 | 5426 | 109 | |
XM_005272620.3 | XP_005272677.1 | 11 - 12 | 5324 | 109 | |
XM_006724536.3 | XP_006724599.1 | 12 - 13 | 6256 | 109 | |
XM_011543929.2 | XP_011542231.1 | 12 - 13 | 4930 | 109 | |
XM_011543930.1 | XP_011542232.1 | 12 - 13 | 4943 | 109 | |
XM_011543931.2 | XP_011542233.1 | 12 - 13 | 5053 | 109 | |
XM_017029615.1 | XP_016885104.1 | 12 - 13 | 6154 | 109 | |
XM_017029616.1 | XP_016885105.1 | 12 - 13 | 5480 | 109 | |
GenBank mRNA | AF317391.1 | - | 12 - 13 | 4879 | 109 |
AK074286.1 | - | 8 - 9 | 3928 | 109 | |
AK095360.1 | - | 7 - 8 | 1631 | 109 | |
AK225581.1 | - | 4 - 5 | 750 | 109 | |
AY316592.1 | - | 12 - 13 | 4981 | 109 | |
BC009675.2 | - | 3 - 4 | 548 | 109 | |
BC114220.1 | - | 11 - 12 | 4937 | 109 |