Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCOR
Entrez Gene ID
54880
Gene Name
BCL6 corepressor
Gene Aliases
ANOP2, MAA2, MCOPS2
Chromosome Location
Chr.X: 40051246 - 40177390 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123383.1 NP_001116855.1 4 - 5 3358 100
NM_001123385.1 NP_001116857.1 4 - 5 3287 100
NM_017745.5 NP_060215.4 4 - 5 3287 100
XM_005272616.1 XP_005272673.1 4 - 5 3419 100
XM_005272618.3 XP_005272675.1 4 - 5 3085 100
XM_006724536.3 XP_006724599.1 4 - 5 4521 100
XM_011543929.2 XP_011542231.1 4 - 5 3195 100
XM_011543930.1 XP_011542232.1 4 - 5 3208 100
XM_011543931.2 XP_011542233.1 4 - 5 3318 100
XM_017029615.1 XP_016885104.1 4 - 5 4521 100
XM_017029616.1 XP_016885105.1 4 - 5 3745 100
GenBank mRNA AF317391.1 - 4 - 5 3246 100
AK056538.1 - 4 - 5 3112 100
AY316592.1 - 4 - 5 3246 100

View Full Product Details