The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321207.1 | NP_001308136.1 | 13 - 14 | 1966 | 142 |
NM_015355.3 | NP_056170.2 | 14 - 15 | 2035 | 142 | |
XM_006721794.3 | XP_006721857.1 | 12 - 13 | 1477 | 142 | |
XM_017024409.1 | XP_016879898.1 | 13 - 14 | 1691 | 142 | |
GenBank mRNA | AK074333.1 | - | 9 - 10 | 1272 | 142 |
AK290014.1 | - | 14 - 15 | 2006 | 142 | |
BC015704.1 | - | 14 - 15 | 1877 | 142 | |
BC018583.1 | - | 13 - 14 | 1909 | 142 | |
D63881.2 | - | 14 - 15 | 1992 | 142 |