Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SUZ12
Entrez Gene ID
23512
Gene Name
SUZ12 polycomb repressive complex 2 subunit
Gene Aliases
CHET9, JJAZ1
Chromosome Location
Chr.17: 31937010 - 32001045 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321207.1 NP_001308136.1 9 - 10 1372 111
NM_015355.3 NP_056170.2 10 - 11 1441 111
XM_006721794.3 XP_006721857.1 8 - 9 883 111
XM_017024409.1 XP_016879898.1 9 - 10 1097 111
GenBank mRNA AK074333.1 - 5 - 6 678 111
AK290014.1 - 10 - 11 1412 111
BC015704.1 - 10 - 11 1283 111
BC018583.1 - 9 - 10 1315 111
D63881.2 - 10 - 11 1398 111

View Full Product Details