Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RHBDL3
Entrez Gene ID
162494
Gene Name
rhomboid like 3
Gene Aliases
RHBDL4, VRHO
Chromosome Location
Chr.17: 32266167 - 32324661 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138328.2 NP_612201.1 2 - 3 150 75
XM_017024272.1 XP_016879761.1 2 - 3 155 75
GenBank mRNA AJ313480.1 - 2 - 3 150 75
BC100975.1 - 2 - 3 149 75
BC100976.1 - 2 - 3 150 75

View Full Product Details