Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NAV2
- Entrez Gene ID
-
89797
- Gene Name
- neuron navigator 2
- Gene Aliases
-
HELAD1, POMFIL2, RAINB1, STEERIN2, UNC53H2
- Chromosome Location
-
Chr.11: 19345200 - 20121601 on Build GRCh38
View Full Product Details