Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCC8
Entrez Gene ID
6833
Gene Name
ATP binding cassette subfamily C member 8
Gene Aliases
ABC36, HHF1, HI, HRINS, MRP8, PHHI, SUR, SUR1, SUR1delta2, TNDM2
Chromosome Location
Chr.11: 17392885 - 17476870 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000352.4 NP_000343.2 37 - 38 4619 59
NM_001287174.1 NP_001274103.1 37 - 38 4622 59
XM_011520331.2 XP_011518633.1 37 - 38 4644 59
XM_017018197.1 XP_016873686.1 37 - 38 4682 59
XM_017018198.1 XP_016873687.1 37 - 38 4679 59
XM_017018199.1 XP_016873688.1 37 - 38 4687 59
XM_017018200.1 XP_016873689.1 37 - 38 4637 59
XM_017018202.1 XP_016873691.1 36 - 37 4483 59
XM_017018204.1 XP_016873693.1 31 - 32 3595 59
GenBank mRNA AF087138.1 - 37 - 38 4585 59
BM313666.1 - 2 - 3 59
L40625.1 - 9 - 10 993 59
L78207.1 - 37 - 38 4588 59
U63421.1 - 37 - 38 4550 59

View Full Product Details