Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WIPI2
Entrez Gene ID
26100
Gene Name
WD repeat domain, phosphoinositide interacting 2
Gene Aliases
ATG18B, Atg21, CGI-50, WIPI-2
Chromosome Location
Chr.7: 5190204 - 5233855 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033518.1 NP_001028690.1 5 - 6 688 83
NM_001033519.1 NP_001028691.1 4 - 5 634 83
NM_001033520.1 NP_001028692.1 2 - 3 527 83
NM_001278299.1 NP_001265228.1 5 - 6 862 83
NM_015610.3 NP_056425.1 5 - 6 688 83
NM_016003.3 NP_057087.2 4 - 5 634 83
XM_006715685.1 XP_006715748.1 3 - 4 520 83
XM_017011952.1 XP_016867441.1 3 - 4 520 83
GenBank mRNA AF151808.1 - 2 - 3 526 83
AK023041.1 - 5 - 6 648 83
AK024279.1 - 4 - 5 613 83
AK315468.1 - 5 - 6 808 83
AL080155.1 - 5 - 6 689 83
AY691426.1 - 2 - 3 458 83
BC004116.2 - 5 - 6 631 83
BC007596.1 - 4 - 5 601 83
BC021012.1 - 4 - 5 553 83
BC021068.1 - 4 - 5 553 83
BC021200.2 - 5 - 6 629 83
CR533530.1 - 5 - 6 472 83

View Full Product Details