Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WIPI2
Entrez Gene ID
26100
Gene Name
WD repeat domain, phosphoinositide interacting 2
Gene Aliases
ATG18B, Atg21, CGI-50, WIPI-2
Chromosome Location
Chr.7: 5190204 - 5233855 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033518.1 NP_001028690.1 12 - 13 1437 129
NM_001033519.1 NP_001028691.1 11 - 12 1383 129
NM_001033520.1 NP_001028692.1 9 - 10 1276 129
XM_017011952.1 XP_016867441.1 10 - 11 1269 129
GenBank mRNA AF151808.1 - 9 - 10 1275 129
AY691425.1 - 11 - 12 1167 129
AY691426.1 - 9 - 10 1207 129
BC021012.1 - 11 - 12 1302 129
BC021068.1 - 11 - 12 1302 129

View Full Product Details