The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033518.1 | NP_001028690.1 | 2 - 3 | 343 | 127 |
NM_015610.3 | NP_056425.1 | 2 - 3 | 343 | 127 | |
GenBank mRNA | AK023041.1 | - | 2 - 3 | 303 | 127 |
AL080155.1 | - | 2 - 3 | 344 | 127 | |
BC004116.2 | - | 2 - 3 | 286 | 127 | |
BC021200.2 | - | 2 - 3 | 284 | 127 | |
CR533530.1 | - | 2 - 3 | 127 | 127 |