The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015254.3 | NP_056069.2 | 2 - 3 | 206 | 144 |
XM_005273458.1 | XP_005273515.1 | 2 - 3 | 206 | 144 | |
XM_011544457.1 | XP_011542759.1 | 2 - 3 | 206 | 144 | |
XM_011544458.1 | XP_011542760.1 | 2 - 3 | 206 | 144 | |
XM_011544459.2 | XP_011542761.1 | 2 - 3 | 191 | 144 | |
XM_011544460.2 | XP_011542762.1 | 2 - 3 | 198 | 144 | |
XM_017013257.1 | XP_016868746.1 | 2 - 3 | 206 | 144 | |
GenBank mRNA | AB014539.3 | - | 2 - 3 | 176 | 144 |
AF279865.1 | - | 2 - 3 | 184 | 144 | |
AK307848.1 | - | 2 - 3 | 185 | 144 | |
BC005977.1 | - | 2 - 3 | 168 | 144 | |
DA068190.1 | - | 2 - 3 | 206 | 144 |