The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037334.1 | NP_001032411.1 | 13 - 14 | 1335 | 133 |
NM_005151.3 | NP_005142.1 | 14 - 15 | 1440 | 133 | |
GenBank mRNA | AK297605.1 | - | 13 - 14 | 1354 | 133 |
AK315438.1 | - | 14 - 15 | 1390 | 133 | |
BC003556.1 | - | 14 - 15 | 1370 | 133 | |
BT007183.1 | - | 14 - 15 | 1224 | 133 | |
U30888.1 | - | 14 - 15 | 1315 | 133 |