The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030649.2 | NP_085152.2 | 16 - 17 | 1486 | 96 |
XM_005244715.2 | XP_005244772.1 | 16 - 17 | 1522 | 96 | |
XM_005244717.3 | XP_005244774.1 | 4 - 5 | 480 | 96 | |
XM_011540606.2 | XP_011538908.1 | 16 - 17 | 1535 | 96 | |
XM_011540607.1 | XP_011538909.1 | 16 - 17 | 3959 | 96 | |
XM_011540608.2 | XP_011538910.1 | 15 - 16 | 1440 | 96 | |
XM_011540609.2 | XP_011538911.1 | 16 - 17 | 1536 | 96 | |
XM_017000233.1 | XP_016855722.1 | 16 - 17 | 1675 | 96 | |
GenBank mRNA | AB051503.1 | - | 15 - 16 | 3151 | 96 |
AF411981.1 | - | 14 - 15 | 1286 | 96 | |
BC004874.2 | - | 1 - 2 | 90 | 96 | |
BC047001.1 | - | 6 - 7 | 938 | 96 | |
BC051194.1 | - | 16 - 17 | 1486 | 96 |