The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030649.2 | NP_085152.2 | 10 - 11 | 829 | 94 |
XM_005244715.2 | XP_005244772.1 | 10 - 11 | 865 | 94 | |
XM_011540606.2 | XP_011538908.1 | 10 - 11 | 878 | 94 | |
XM_011540607.1 | XP_011538909.1 | 10 - 11 | 3302 | 94 | |
XM_011540608.2 | XP_011538910.1 | 9 - 10 | 783 | 94 | |
XM_011540609.2 | XP_011538911.1 | 10 - 11 | 879 | 94 | |
XM_017000233.1 | XP_016855722.1 | 10 - 11 | 1018 | 94 | |
GenBank mRNA | AB051503.1 | - | 9 - 10 | 2494 | 94 |
AF411981.1 | - | 8 - 9 | 629 | 94 | |
BC051194.1 | - | 10 - 11 | 829 | 94 |