Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPTBN4
Entrez Gene ID
57731
Gene Name
spectrin beta, non-erythrocytic 4
Gene Aliases
QV, SPNB4, SPTBN3
Chromosome Location
Chr.19: 40467153 - 40576459 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020971.2 NP_066022.2 16 - 17 3757 102
XM_011527173.2 XP_011525475.1 16 - 17 3823 102
XM_017027049.1 XP_016882538.1 16 - 17 4010 102
XM_017027050.1 XP_016882539.1 14 - 15 3392 102
XM_017027051.1 XP_016882540.1 16 - 17 3823 102
XM_017027052.1 XP_016882541.1 16 - 17 3823 102
GenBank mRNA AB046862.1 - 7 - 8 2500 102
AF082075.1 - 16 - 17 3818 102
AF311855.1 - 16 - 17 3757 102
AF324063.1 - 16 - 17 3741 102
AF324064.1 - 16 - 17 3741 102
AY004226.1 - 15 - 16 3640 102
AY004227.1 - 15 - 16 3640 102

View Full Product Details