The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032012.3 | NP_114401.2 | 1 - 2 | 612 | 65 |
XM_011518446.2 | XP_011516748.1 | 1 - 2 | 630 | 65 | |
XM_011518447.1 | XP_011516749.1 | 1 - 2 | 612 | 65 | |
XM_011518449.1 | XP_011516751.1 | 1 - 2 | 612 | 65 | |
XM_011518451.1 | XP_011516753.1 | 1 - 2 | 612 | 65 | |
XM_011518452.1 | XP_011516754.1 | 1 - 2 | 612 | 65 | |
XM_017014571.1 | XP_016870060.1 | 1 - 2 | 612 | 65 | |
XM_017014572.1 | XP_016870061.1 | 1 - 2 | 612 | 65 | |
GenBank mRNA | AA465408.1 | - | 4 - 5 | 65 | |
AF153415.1 | - | 1 - 2 | 612 | 65 | |
AK299952.1 | - | 1 - 2 | 594 | 65 | |
BE675782.1 | - | 1 - 2 | 298 | 65 | |
BP351493.1 | - | 1 - 2 | 133 | 65 |