The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081560.2 | NP_001075029.1 | 14 - 15 | 1924 | 76 |
NM_001081562.2 | NP_001075031.1 | 14 - 15 | 1920 | 76 | |
NM_001081563.2 | NP_001075032.1 | 13 - 14 | 2308 | 76 | |
NM_001288764.1 | NP_001275693.1 | 15 - 16 | 1998 | 76 | |
NM_001288765.1 | NP_001275694.1 | 12 - 13 | 1575 | 76 | |
NM_004409.4 | NP_004400.4 | 14 - 15 | 1939 | 76 | |
GenBank mRNA | AB209363.1 | - | 14 - 15 | 2763 | 76 |
AK297299.1 | - | 12 - 13 | 1526 | 76 | |
BC026328.1 | - | 15 - 16 | 1993 | 76 | |
BC062553.1 | - | 14 - 15 | 1860 | 76 | |
KJ534826.1 | - | 13 - 14 | 1764 | 76 | |
KJ534827.1 | - | 13 - 14 | 1764 | 76 | |
KJ534828.1 | - | 13 - 14 | 1749 | 76 | |
L19268.1 | - | 14 - 15 | 1846 | 76 | |
M87312.1 | - | 13 - 14 | 2309 | 76 | |
S72883.1 | - | 14 - 15 | 1751 | 76 | |
S86455.1 | - | 1 - 2 | 56 | 76 | |
U46546.1 | - | 2 - 3 | 105 | 76 |