Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR3
Entrez Gene ID
10885
Gene Name
WD repeat domain 3
Gene Aliases
DIP2, UTP12
Chromosome Location
Chr.1: 117929749 - 117960426 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006784.2 NP_006775.1 16 - 17 1873 63
GenBank mRNA AB209201.1 - 4 - 5 378 63
AF083217.1 - 16 - 17 1854 63
AK022858.1 - 8 - 9 878 63
AK292438.1 - 16 - 17 1868 63

View Full Product Details