Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR3
Entrez Gene ID
10885
Gene Name
WD repeat domain 3
Gene Aliases
DIP2, UTP12
Chromosome Location
Chr.1: 117929749 - 117960426 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006784.2 NP_006775.1 4 - 5 564 90
GenBank mRNA AF083217.1 - 4 - 5 545 90
AK292438.1 - 4 - 5 559 90
BC058836.1 - 5 - 6 603 90

View Full Product Details