Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D8B
Entrez Gene ID
54885
Gene Name
TBC1 domain family member 8B
Gene Aliases
GRAMD8B
Chromosome Location
Chr.X: 106802689 - 106876150 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017752.2 NP_060222.2 12 - 13 2281 107
XM_006724669.1 XP_006724732.1 11 - 12 2131 107
XM_006724670.3 XP_006724733.1 12 - 13 2095 107
GenBank mRNA AB449890.1 - 12 - 13 2116 107
AK091248.1 - 3 - 4 598 107

View Full Product Details