The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297703.1 | NP_001284632.1 | 4 - 5 | 797 | 89 |
NM_001321417.1 | NP_001308346.1 | 5 - 6 | 1056 | 89 | |
NM_031941.3 | NP_114147.2 | 5 - 6 | 885 | 89 | |
NR_135632.1 | 5 - 6 | 946 | 89 | ||
XM_011528351.2 | XP_011526653.1 | 5 - 6 | 946 | 89 | |
GenBank mRNA | AB026290.1 | - | 5 - 6 | 864 | 89 |
AB040046.1 | - | 4 - 5 | 1875 | 89 | |
AK096028.1 | - | 5 - 6 | 878 | 89 | |
AK292031.1 | - | 5 - 6 | 914 | 89 | |
AK300617.1 | - | 4 - 5 | 725 | 89 | |
AK315239.1 | - | 5 - 6 | 888 | 89 | |
BC027910.1 | - | 5 - 6 | 885 | 89 |