The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020812.3 | NP_065863.2 | 17 - 18 | 2027 | 95 |
XM_005260000.2 | XP_005260057.1 | 17 - 18 | 2036 | 95 | |
XM_005260001.2 | XP_005260058.1 | 17 - 18 | 2036 | 95 | |
XM_011528150.1 | XP_011526452.1 | 17 - 18 | 2069 | 95 | |
XM_011528151.1 | XP_011526453.1 | 17 - 18 | 2069 | 95 | |
XM_011528152.1 | XP_011526454.1 | 17 - 18 | 2069 | 95 | |
GenBank mRNA | AB037816.2 | - | 17 - 18 | 1987 | 95 |
AK295664.1 | - | 2 - 3 | 186 | 95 | |
BC146786.1 | - | 17 - 18 | 1987 | 95 | |
DC333132.1 | - | 2 - 3 | 186 | 95 |