Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- DST
- Entrez Gene ID
-
667
- Gene Name
- dystonin
- Gene Aliases
-
BP240, BPA, BPAG1, CATX-15, CATX15, D6S1101, DMH, DT, EBSB2, HSAN6, MACF2
- Chromosome Location
-
Chr.6: 56457987 - 56954671 on Build GRCh38
View Full Product Details