Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DST
Entrez Gene ID
667
Gene Name
dystonin
Gene Aliases
BP240, BPA, BPAG1, CATX-15, CATX15, D6S1101, DMH, DT, EBSB2, HSAN6, MACF2
Chromosome Location
Chr.6: 56457987 - 56954671 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144769.2 NP_001138241.1 25 - 26 3436 81
NM_001144770.1 NP_001138242.1 23 - 24 3107 81
NM_001723.5 NP_001714.1 12 - 13 1991 81
NM_015548.4 NP_056363.2 12 - 13 1991 81
NM_183380.3 NP_899236.1 22 - 23 3381 81
XM_005249310.3 XP_005249367.1 26 - 27 3530 81
XM_005249315.3 XP_005249372.1 25 - 26 3477 81
XM_005249316.2 XP_005249373.1 18 - 19 3753 81
XM_005249318.2 XP_005249375.1 24 - 25 3191 81
XM_005249319.2 XP_005249376.1 23 - 24 3092 81
XM_005249320.4 XP_005249377.1 23 - 24 3148 81
XM_005249322.4 XP_005249379.1 23 - 24 3043 81
XM_005249323.3 XP_005249380.1 22 - 23 3376 81
XM_005249324.2 XP_005249381.1 12 - 13 1988 81
XM_011514824.2 XP_011513126.1 25 - 26 3441 81
XM_011514825.2 XP_011513127.1 25 - 26 3437 81
XM_011514826.2 XP_011513128.1 22 - 23 3376 81
XM_017011205.1 XP_016866694.1 26 - 27 3540 81
XM_017011206.1 XP_016866695.1 26 - 27 3540 81
XM_017011207.1 XP_016866696.1 26 - 27 3477 81
XM_017011208.1 XP_016866697.1 26 - 27 3540 81
XM_017011209.1 XP_016866698.1 26 - 27 3540 81
XM_017011210.1 XP_016866699.1 26 - 27 3540 81
XM_017011211.1 XP_016866700.1 26 - 27 3540 81
XM_017011212.1 XP_016866701.1 26 - 27 3540 81
XM_017011213.1 XP_016866702.1 26 - 27 3540 81
XM_017011214.1 XP_016866703.1 26 - 27 3540 81
XM_017011215.1 XP_016866704.1 26 - 27 3540 81
XM_017011216.1 XP_016866705.1 26 - 27 3540 81
XM_017011217.1 XP_016866706.1 23 - 24 3121 81
XM_017011219.1 XP_016866708.1 26 - 27 3536 81
XM_017011220.1 XP_016866709.1 25 - 26 3479 81
XM_017011221.1 XP_016866710.1 26 - 27 3536 81
XM_017011222.1 XP_016866711.1 18 - 19 3695 81
XM_017011223.1 XP_016866712.1 26 - 27 3536 81
XM_017011224.1 XP_016866713.1 22 - 23 3376 81
XM_017011225.1 XP_016866714.1 26 - 27 3550 81
GenBank mRNA AF400226.1 - 12 - 13 1883 81
AF400227.1 - 12 - 13 1883 81
AK299898.1 - 23 - 24 3107 81
AU117550.1 - 2 - 3 254 81
BC065536.1 - 12 - 13 1955 81
L11690.1 - 12 - 13 1985 81
M63618.1 - 2 - 3 222 81
M69225.1 - 12 - 13 1921 81

View Full Product Details