Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DST
Entrez Gene ID
667
Gene Name
dystonin
Gene Aliases
BP240, BPA, BPAG1, CATX-15, CATX15, D6S1101, DMH, DT, EBSB2, HSAN6, MACF2
Chromosome Location
Chr.6: 56457987 - 56954671 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144769.2 NP_001138241.1 20 - 21 2792 110
NM_001144770.1 NP_001138242.1 18 - 19 2463 110
NM_001723.5 NP_001714.1 7 - 8 1347 110
NM_015548.4 NP_056363.2 7 - 8 1347 110
NM_183380.3 NP_899236.1 17 - 18 2737 110
XM_005249310.3 XP_005249367.1 21 - 22 2886 110
XM_005249315.3 XP_005249372.1 20 - 21 2833 110
XM_005249316.2 XP_005249373.1 13 - 14 3109 110
XM_005249318.2 XP_005249375.1 19 - 20 2547 110
XM_005249319.2 XP_005249376.1 18 - 19 2448 110
XM_005249320.4 XP_005249377.1 18 - 19 2504 110
XM_005249322.4 XP_005249379.1 18 - 19 2399 110
XM_005249323.3 XP_005249380.1 17 - 18 2732 110
XM_005249324.2 XP_005249381.1 7 - 8 1344 110
XM_011514824.2 XP_011513126.1 20 - 21 2797 110
XM_011514825.2 XP_011513127.1 20 - 21 2793 110
XM_011514826.2 XP_011513128.1 17 - 18 2732 110
XM_017011205.1 XP_016866694.1 21 - 22 2896 110
XM_017011206.1 XP_016866695.1 21 - 22 2896 110
XM_017011207.1 XP_016866696.1 21 - 22 2833 110
XM_017011208.1 XP_016866697.1 21 - 22 2896 110
XM_017011209.1 XP_016866698.1 21 - 22 2896 110
XM_017011210.1 XP_016866699.1 21 - 22 2896 110
XM_017011211.1 XP_016866700.1 21 - 22 2896 110
XM_017011212.1 XP_016866701.1 21 - 22 2896 110
XM_017011213.1 XP_016866702.1 21 - 22 2896 110
XM_017011214.1 XP_016866703.1 21 - 22 2896 110
XM_017011215.1 XP_016866704.1 21 - 22 2896 110
XM_017011216.1 XP_016866705.1 21 - 22 2896 110
XM_017011217.1 XP_016866706.1 18 - 19 2477 110
XM_017011219.1 XP_016866708.1 21 - 22 2892 110
XM_017011220.1 XP_016866709.1 20 - 21 2835 110
XM_017011221.1 XP_016866710.1 21 - 22 2892 110
XM_017011222.1 XP_016866711.1 13 - 14 3051 110
XM_017011223.1 XP_016866712.1 21 - 22 2892 110
XM_017011224.1 XP_016866713.1 17 - 18 2732 110
XM_017011225.1 XP_016866714.1 21 - 22 2906 110
GenBank mRNA AF400226.1 - 7 - 8 1239 110
AF400227.1 - 7 - 8 1239 110
AK294830.1 - 17 - 18 2345 110
AK295864.1 - 13 - 14 2719 110
AK299898.1 - 18 - 19 2463 110
BC065536.1 - 7 - 8 1311 110
L11690.1 - 7 - 8 1341 110
M69225.1 - 7 - 8 1277 110

View Full Product Details