Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DROSHA
Entrez Gene ID
29102
Gene Name
drosha ribonuclease III
Gene Aliases
ETOHI2, HSA242976, RANSE3L, RN3, RNASE3L, RNASEN
Chromosome Location
Chr.5: 31400494 - 31532175 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100412.1 NP_001093882.1 21 - 22 3139 77
NM_013235.4 NP_037367.3 21 - 22 3174 77
XM_005248291.3 XP_005248348.1 22 - 23 3250 77
XM_005248292.3 XP_005248349.1 22 - 23 3226 77
XM_005248293.3 XP_005248350.1 21 - 22 3157 77
XM_005248294.3 XP_005248351.1 20 - 21 3046 77
XM_011514033.2 XP_011512335.1 22 - 23 3250 77
XM_017009399.1 XP_016864888.1 21 - 22 3133 77
XM_017009400.1 XP_016864889.1 21 - 22 3115 77
XM_017009401.1 XP_016864890.1 20 - 21 3022 77
XM_017009402.1 XP_016864891.1 10 - 11 1265 77
GenBank mRNA AB209501.1 - 14 - 15 10769 77
AF116910.1 - 6 - 7 703 77
AF189011.1 - 21 - 22 3060 77
AK001121.1 - 10 - 11 1009 77
AK026563.1 - 1 - 2 61 77
AK308093.1 - 20 - 21 2948 77
BC024261.1 - 9 - 10 935 77
BC041162.1 - 8 - 9 860 77
BC054003.1 - 20 - 21 2951 77
BX647724.1 - 21 - 22 3042 77

View Full Product Details