Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIAA0101
Entrez Gene ID
9768
Gene Name
KIAA0101
Gene Aliases
L5, NS5ATP9, OEATC, OEATC-1, OEATC1, PAF, PAF15, p15(PAF), p15/PAF, p15PAF
Chromosome Location
Chr.15: 64364994 - 64387687 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_109934.1 1 - 2 246 131
GenBank mRNA DT220867.1 - 1 - 2 243 131

View Full Product Details