Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MLLT1
Entrez Gene ID
4298
Gene Name
MLLT1, super elongation complex subunit
Gene Aliases
ENL, LTG19, YEATS1
Chromosome Location
Chr.19: 6210381 - 6279948 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005934.3 NP_005925.2 12 - 12 3638 87
XM_011528021.1 XP_011526323.1 12 - 12 3612 87
XM_011528022.1 XP_011526324.1 12 - 12 3495 87
XM_011528023.1 XP_011526325.1 11 - 11 3486 87
XM_017026819.1 XP_016882308.1 12 - 12 4194 87
GenBank mRNA AL365410.1 - 1 - 1 983 87
BC047008.1 - 1 - 1 1173 87
BC084564.1 - 1 - 1 1144 87

View Full Product Details