The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024753.4 | NP_079029.3 | 19 - 20 | 2706 | 134 |
XM_006712761.1 | XP_006712824.1 | 19 - 20 | 2649 | 134 | |
XM_011511871.2 | XP_011510173.1 | 14 - 15 | 5951 | 134 | |
XM_011511872.2 | XP_011510174.1 | 19 - 20 | 2649 | 134 | |
XM_017004967.1 | XP_016860456.1 | 19 - 20 | 2649 | 134 | |
XM_017004968.1 | XP_016860457.1 | 14 - 15 | 4197 | 134 | |
XM_017004969.1 | XP_016860458.1 | 17 - 18 | 2366 | 134 | |
GenBank mRNA | AK096451.1 | - | 3 - 4 | 333 | 134 |
AK292942.1 | - | 19 - 20 | 2649 | 134 | |
BC055424.1 | - | 19 - 20 | 2706 | 134 | |
BX647903.1 | - | 5 - 6 | 457 | 134 |