The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256090.1 | NP_001243019.1 | 8 - 9 | 779 | 67 |
NM_001256091.1 | NP_001243020.1 | 7 - 8 | 704 | 67 | |
NM_001256092.1 | NP_001243021.1 | 6 - 7 | 647 | 67 | |
NM_001256093.1 | NP_001243022.1 | 6 - 7 | 647 | 67 | |
NM_001256094.1 | NP_001243023.1 | 7 - 8 | 744 | 67 | |
NM_001880.3 | NP_001871.2 | 7 - 8 | 744 | 67 | |
NR_045768.1 | 8 - 9 | 779 | 67 | ||
NR_045769.1 | 8 - 9 | 779 | 67 | ||
NR_045770.1 | 8 - 9 | 779 | 67 | ||
NR_045771.1 | 7 - 8 | 744 | 67 | ||
NR_045772.1 | 7 - 8 | 682 | 67 | ||
NR_045773.1 | 6 - 7 | 647 | 67 | ||
NR_045774.1 | 7 - 8 | 744 | 67 | ||
GenBank mRNA | AK128731.1 | - | 7 - 8 | 644 | 67 |
BC026175.1 | - | 7 - 8 | 711 | 67 | |
BC107698.1 | - | 6 - 7 | 614 | 67 | |
BC130335.1 | - | 6 - 7 | 510 | 67 | |
BC130337.1 | - | 6 - 7 | 510 | 67 | |
BX648469.1 | - | 6 - 7 | 534 | 67 | |
DQ003036.1 | - | 8 - 9 | 742 | 67 | |
DQ003037.1 | - | 7 - 8 | 707 | 67 | |
DQ003039.1 | - | 7 - 8 | 707 | 67 | |
DQ003040.1 | - | 7 - 8 | 645 | 67 | |
DQ003041.1 | - | 8 - 9 | 742 | 67 | |
DQ003042.1 | - | 7 - 8 | 645 | 67 | |
DQ003043.1 | - | 8 - 9 | 742 | 67 | |
DQ003044.1 | - | 8 - 9 | 742 | 67 | |
DQ003045.1 | - | 6 - 7 | 610 | 67 | |
DQ003046.1 | - | 6 - 7 | 610 | 67 | |
DQ003047.1 | - | 7 - 8 | 707 | 67 | |
DQ003048.1 | - | 6 - 7 | 610 | 67 | |
DQ003049.1 | - | 7 - 8 | 667 | 67 | |
X15875.1 | - | 5 - 6 | 471 | 67 |