The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005941.4 | NP_005932.2 | 3 - 4 | 686 | 70 |
GenBank mRNA | AB009303.1 | - | 3 - 4 | 516 | 70 |
AK314274.1 | - | 3 - 4 | 686 | 70 | |
AL832595.1 | - | 4 - 5 | 725 | 70 | |
BC069500.1 | - | 3 - 4 | 473 | 70 | |
BC075004.2 | - | 3 - 4 | 430 | 70 | |
BC075005.2 | - | 3 - 4 | 430 | 70 | |
D83646.1 | - | 3 - 4 | 404 | 70 | |
D83647.1 | - | 3 - 4 | 404 | 70 | |
D85511.1 | - | 3 - 4 | 439 | 70 |