The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001848.2 | NP_001839.2 | 34 - 35 | 2573 | 85 |
GenBank mRNA | AK298968.1 | - | 17 - 18 | 1281 | 85 |
BC005159.2 | - | 8 - 9 | 723 | 85 | |
BC022236.1 | - | 5 - 6 | 705 | 85 | |
BC052575.1 | - | 34 - 35 | 2553 | 85 | |
X15880.1 | - | 7 - 8 | 695 | 85 |