The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164615.1 | NP_001158087.1 | 4 - 5 | 1683 | 62 |
NM_001164616.1 | NP_001158088.1 | 5 - 6 | 1595 | 62 | |
NM_001317796.1 | NP_001304725.1 | 3 - 4 | 940 | 62 | |
NM_003506.3 | NP_003497.2 | 4 - 5 | 1710 | 62 | |
GenBank mRNA | AB012911.1 | - | 4 - 5 | 1651 | 62 |
AF072873.1 | - | 4 - 5 | 1673 | 62 | |
AK296824.1 | - | 3 - 4 | 895 | 62 | |
AK299341.1 | - | 5 - 6 | 1561 | 62 | |
AK304092.1 | - | 5 - 6 | 1718 | 62 | |
AK313789.1 | - | 4 - 5 | 1654 | 62 | |
AK316544.1 | - | 5 - 6 | 1528 | 62 | |
BC060836.1 | - | 4 - 5 | 1666 | 62 | |
BX640609.1 | - | 4 - 5 | 1677 | 62 |