The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191057.3 | NP_001177986.1 | 16 - 17 | 2366 | 121 |
NM_001191058.3 | NP_001177987.2 | 17 - 18 | 2746 | 121 | |
NM_001191059.3 | NP_001177988.1 | 17 - 18 | 2357 | 121 | |
NM_001322055.1 | NP_001308984.1 | 17 - 18 | 2592 | 121 | |
XM_017012264.1 | XP_016867753.1 | 17 - 18 | 2197 | 121 | |
XM_017012265.1 | XP_016867754.1 | 17 - 18 | 2048 | 121 | |
XM_017012267.1 | XP_016867756.1 | 17 - 18 | 2102 | 121 | |
GenBank mRNA | AK056170.1 | - | 16 - 17 | 2353 | 121 |
AK091734.1 | - | 17 - 18 | 2671 | 121 | |
AK094180.1 | - | 17 - 18 | 2289 | 121 | |
U40372.1 | - | 12 - 13 | 1463 | 121 |