Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UPB1
Entrez Gene ID
51733
Gene Name
beta-ureidopropionase 1
Gene Aliases
BUP1
Chromosome Location
Chr.22: 24495060 - 24528681 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016327.2 NP_057411.1 3 - 4 490 85
GenBank mRNA AB013885.1 - 3 - 4 429 85
AF163312.1 - 3 - 4 430 85
AK024277.1 - 3 - 4 1758 85
AK314727.1 - 3 - 4 626 85
BC033388.1 - 3 - 4 387 85
BC131703.1 - 3 - 4 443 85
BC139843.1 - 3 - 4 388 85
CR456375.1 - 3 - 4 490 85

View Full Product Details