The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024896.2 | NP_079172.2 | 2 - 3 | 699 | 86 |
XM_005251587.1 | XP_005251644.1 | 2 - 3 | 670 | 86 | |
XM_017015139.1 | XP_016870628.1 | 3 - 4 | 492 | 86 | |
GenBank mRNA | AB058718.2 | - | 2 - 3 | 576 | 86 |
AK093217.1 | - | 1 - 2 | 66 | 86 | |
AK127218.1 | - | 2 - 3 | 688 | 86 | |
BC136771.1 | - | 2 - 3 | 673 | 86 | |
BC136773.1 | - | 2 - 3 | 673 | 86 | |
BC144404.1 | - | 2 - 3 | 673 | 86 |