Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CADPS2
Entrez Gene ID
93664
Gene Name
calcium dependent secretion activator 2
Gene Aliases
CAPS2
Chromosome Location
Chr.7: 122318424 - 122886759 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009571.3 NP_001009571.2 6 - 7 1642 93
NM_001167940.1 NP_001161412.1 6 - 7 1642 93
NM_017954.10 NP_060424.9 6 - 7 1642 93
XM_005250695.4 XP_005250752.1 6 - 7 1371 93
XM_005250696.4 XP_005250753.1 6 - 7 1371 93
XM_005250697.4 XP_005250754.1 6 - 7 1371 93
XM_005250698.4 XP_005250755.1 6 - 7 1343 93
XM_005250699.4 XP_005250756.1 6 - 7 1371 93
XM_005250700.4 XP_005250757.1 6 - 7 1371 93
XM_005250701.4 XP_005250758.1 6 - 7 1371 93
XM_005250702.4 XP_005250759.1 6 - 7 1371 93
XM_005250703.4 XP_005250760.1 6 - 7 1371 93
XM_005250704.4 XP_005250761.1 6 - 7 1371 93
XM_005250705.4 XP_005250762.1 6 - 7 1371 93
XM_005250706.4 XP_005250763.1 6 - 7 1371 93
XM_005250707.4 XP_005250764.1 6 - 7 1371 93
XM_005250708.3 XP_005250765.1 6 - 7 1371 93
XM_011516695.2 XP_011514997.1 6 - 7 1343 93
XM_011516696.2 XP_011514998.1 6 - 7 1371 93
XM_011516697.2 XP_011514999.1 6 - 7 1371 93
XM_017012794.1 XP_016868283.1 6 - 7 1371 93
XM_017012795.1 XP_016868284.1 6 - 7 1371 93
XM_017012796.1 XP_016868285.1 7 - 8 986 93
XM_017012797.1 XP_016868286.1 7 - 8 1052 93
XM_017012798.1 XP_016868287.1 6 - 7 1522 93
XM_017012799.1 XP_016868288.1 7 - 8 1423 93
XM_017012800.1 XP_016868289.1 3 - 4 379 93
GenBank mRNA AB046811.1 - 4 - 5 511 93
AF401638.1 - 6 - 7 1311 93
AK025672.1 - 2 - 3 165 93
AK126518.1 - 7 - 8 1244 93
AL833058.1 - 3 - 4 437 93
AY264289.1 - 6 - 7 1246 93
BC136601.1 - 6 - 7 1341 93
BC144278.1 - 6 - 7 1341 93
BC144279.1 - 6 - 7 1341 93
KJ534798.1 - 6 - 7 1220 93
KJ534800.1 - 6 - 7 1220 93
KJ534801.1 - 6 - 7 1220 93
KJ534803.1 - 6 - 7 1220 93
KJ534804.1 - 6 - 7 1220 93
KJ534805.1 - 6 - 7 1220 93

View Full Product Details