The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144982.4 | NP_659419.3 | 34 - 35 | 6189 | 142 |
GenBank mRNA | AB011118.2 | - | 33 - 34 | 6727 | 142 |
AK125035.1 | - | 25 - 26 | 3643 | 142 | |
BX647709.1 | - | 34 - 35 | 6069 | 142 |