The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003406.1 | NP_001003406.1 | 30 - 31 | 4950 | 101 |
NM_021096.3 | NP_066919.2 | 31 - 32 | 5055 | 101 | |
XM_017029035.1 | XP_016884524.1 | 21 - 22 | 8325 | 101 | |
XM_017029036.1 | XP_016884525.1 | 21 - 22 | 8325 | 101 | |
XM_017029037.1 | XP_016884526.1 | 21 - 22 | 8326 | 101 | |
GenBank mRNA | AB032946.1 | - | 12 - 13 | 1459 | 101 |
AF129133.1 | - | 31 - 32 | 5055 | 101 | |
AF142567.1 | - | 31 - 32 | 5055 | 101 | |
AF211189.1 | - | 30 - 31 | 4950 | 101 | |
AF393329.1 | - | 30 - 31 | 4950 | 101 |