The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130466.3 | NP_569733.2 | 15 - 16 | 2234 | 98 |
NM_183415.2 | NP_904324.1 | 15 - 16 | 1889 | 98 | |
XM_005253987.2 | XP_005254044.1 | 15 - 16 | 2114 | 98 | |
XM_006719681.3 | XP_006719744.1 | 15 - 16 | 2447 | 98 | |
XM_006719682.2 | XP_006719745.1 | 15 - 16 | 2447 | 98 | |
XM_011538959.2 | XP_011537261.1 | 14 - 15 | 1996 | 98 | |
XM_011538961.1 | XP_011537263.1 | 15 - 16 | 2447 | 98 | |
XM_017020195.1 | XP_016875684.1 | 15 - 16 | 2657 | 98 | |
GenBank mRNA | AF130072.1 | - | 13 - 14 | 1626 | 98 |
AF251046.2 | - | 15 - 16 | 2204 | 98 | |
AK307385.1 | - | 14 - 15 | 1768 | 98 | |
AL096740.1 | - | 15 - 16 | 2203 | 98 | |
BC032301.1 | - | 15 - 16 | 1867 | 98 | |
BC141880.1 | - | 15 - 16 | 2112 | 98 | |
BC144684.1 | - | 15 - 16 | 2210 | 98 | |
BC150637.1 | - | 15 - 16 | 1865 | 98 | |
BX537403.1 | - | 15 - 16 | 1879 | 98 |