The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127460.2 | NP_001120932.1 | 2 - 3 | 660 | 74 |
NM_001166279.1 | NP_001159751.1 | 2 - 3 | 589 | 74 | |
NM_014871.4 | NP_055686.3 | 2 - 3 | 589 | 74 | |
GenBank mRNA | AB014610.2 | - | 2 - 3 | 486 | 74 |
AB107585.1 | - | 1 - 2 | 283 | 74 | |
BC024043.1 | - | 2 - 3 | 502 | 74 | |
BC094885.1 | - | 2 - 3 | 657 | 74 | |
BX648106.1 | - | 2 - 3 | 577 | 74 |