The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001923.4 | NP_001914.3 | 10 - 11 | 1462 | 75 |
GenBank mRNA | AJ002955.1 | - | 10 - 11 | 1327 | 75 |
AK294493.1 | - | 12 - 13 | 1591 | 75 | |
AK299650.1 | - | 9 - 10 | 911 | 75 | |
AK302924.1 | - | 8 - 9 | 860 | 75 | |
AK312436.1 | - | 10 - 11 | 1284 | 75 | |
AL831958.1 | - | 10 - 11 | 1444 | 75 | |
BC011686.2 | - | 10 - 11 | 1291 | 75 | |
BC021044.1 | - | 10 - 11 | 1279 | 75 | |
BC050530.1 | - | 10 - 11 | 1355 | 75 | |
BC051764.2 | - | 10 - 11 | 1345 | 75 | |
L40326.1 | - | 10 - 11 | 1256 | 75 | |
U18299.1 | - | 10 - 11 | 1318 | 75 |