The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001923.4 | NP_001914.3 | 22 - 23 | 3078 | 73 |
GenBank mRNA | AJ002955.1 | - | 22 - 23 | 2943 | 73 |
AK294341.1 | - | 6 - 7 | 852 | 73 | |
AK294493.1 | - | 24 - 25 | 3207 | 73 | |
AK299650.1 | - | 21 - 22 | 2527 | 73 | |
AK303032.1 | - | 16 - 17 | 1960 | 73 | |
AK312436.1 | - | 22 - 23 | 2900 | 73 | |
BC011686.2 | - | 22 - 23 | 2907 | 73 | |
BC021044.1 | - | 22 - 23 | 2895 | 73 | |
BC032080.1 | - | 8 - 9 | 1007 | 73 | |
BC050530.1 | - | 22 - 23 | 2971 | 73 | |
BC051764.2 | - | 22 - 23 | 2961 | 73 | |
L40326.1 | - | 22 - 23 | 2872 | 73 | |
U18299.1 | - | 22 - 23 | 2934 | 73 | |
U32986.1 | - | 22 - 23 | 2943 | 73 |